CAS RN 65726-24-1
CAS RN 65726-24-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Nabumetone EP Impurity C, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
2-Hydroxy Nabumetone, TRC
CAS: 65726-24-1 Molecular Formula: C15 H18 O2 Molecular Weight (g/mol): 230.3 Synonym: (2RS)-4-(6-Methoxynaphthalen-2-yl)butan-2-ol,Nabumetone Imp. C (EP) IUPAC Name: 4-(6-methoxynaphthalen-2-yl)butan-2-ol SMILES: COc1ccc2cc(CCC(C)O)ccc2c1