missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-6-methylnicotinic acid, 98+%

Product Code. 11443804
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443804 25 g 25g
11433804 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11443804 Supplier Thermo Scientific Alfa Aesar Supplier No. A11842.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38116-61-9 | C7H7NO3 | 153.137 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38116-61-9
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00006272
InChI Key XRIHTJYXIHOBDQ-UHFFFAOYSA-N
Synonym 2-hydroxy-6-methylnicotinic acid, 2-hydroxy-6-methylpyridine-3-carboxylic acid, 6-methyl-2-oxo-1,2-dihydropyridine-3-carboxylic acid, chembl50725, 3-pyridinecarboxylic acid, 1,2-dihydro-6-methyl-2-oxo, 2-hydroxy-6-methyl nicotinic acid, 2-hydroxy-6-methyl-nicotinic acid, 6-methyl-2-oxo-1,2-dihydro-pyridine-3-carboxylic acid, 6-methyl-2-oxohydropyridine-3-carboxylic acid, 2-hydroxy-6-methylnicotinicacid
PubChem CID 94960
IUPAC Name 6-methyl-2-oxo-1H-pyridine-3-carboxylic acid
SMILES CC1=CC=C(C(=O)N1)C(=O)O

Specifications

Melting Point 233°C to 238°C
Quantity 25 g
Beilstein 129164
Formula Weight 153.14
Percent Purity ≥98%
Chemical Name or Material 2-Hydroxy-6-methylnicotinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.