CAS RN 38116-61-9
CAS RN 38116-61-9
IUPAC Name:
6-methyl-2-oxo-1,2-dihydropyridine-3-carboxylate
Synonyms:
6-methyl-2-oxo-1H-pyridine-3-carboxylate
Molecular Weight (g/mol):
152.13
Molecular Formula:
C7H6NO3
InChi Key:
XRIHTJYXIHOBDQ-UHFFFAOYSA-M
SMILES:
CC1=CC=C(C([O-])=O)C(=O)N1
1
–
1
of
1
results
1
2-Hydroxy-6-methylnicotinic acid, 98+%
CAS: 38116-61-9 Molecular Formula: C7H7NO3 Molecular Weight (g/mol): 153.137 MDL Number: MFCD00006272 InChI Key: XRIHTJYXIHOBDQ-UHFFFAOYSA-N Synonym: 2-hydroxy-6-methylnicotinic acid,2-hydroxy-6-methylpyridine-3-carboxylic acid,6-methyl-2-oxo-1,2-dihydropyridine-3-carboxylic acid,chembl50725,3-pyridinecarboxylic acid, 1,2-dihydro-6-methyl-2-oxo,2-hydroxy-6-methyl nicotinic acid,2-hydroxy-6-methyl-nicotinic acid,6-methyl-2-oxo-1,2-dihydro-pyridine-3-carboxylic acid,6-methyl-2-oxohydropyridine-3-carboxylic acid,2-hydroxy-6-methylnicotinicacid PubChem CID: 94960 IUPAC Name: 6-methyl-2-oxo-1H-pyridine-3-carboxylic acid SMILES: CC1=CC=C(C(=O)N1)C(=O)O