missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-Amino-N-(2-carboxy-4-aminophenyl)benzoic Acid, TRC

Product Code. 30399543
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399543 1 mg 1mg
30407834 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30399543 Supplier Toronto Research Chemicals Supplier No. TRCH7680001MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1797983-23-3
Molecular Formula C14 H12 N2 O5
Molecular Weight (g/mol) 288.26
Synonym 5-Amino-2-((3-carboxy-4-hydroxyphenyl)amino)benzoic Acid, N-(2-Carboxy-4-aminophenyl)-5-aminosalicylic Acid, 2-Hydroxy-5-[(2-carboxy-4-aminophenyl)amino]benzoic Acid
IUPAC Name 5-amino-2-(3-carboxy-4-hydroxyanilino)benzoic acid
SMILES Nc1ccc(Nc2ccc(O)c(c2)C(=O)O)c(c1)C(=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C14H12N2O5/c15-7-1-3-11(9(5-7)13(18)19)16-8-2-4-12(17)10(6-8)14(20)21/h1-6,16-17H,15H2,(H,18,19)(H,20,21)
Formula Weight 288.0746
Chemical Name or Material 2-Hydroxy-5-[(2-carboxy-4-aminophenyl)amino]benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.