CAS RN 1797983-23-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Hydroxy-5-Amino-N-(2-carboxy-4-aminophenyl)benzoic Acid, TRC
CAS: 1797983-23-3 Molecular Formula: C14 H12 N2 O5 Molecular Weight (g/mol): 288.26 Synonym: 5-Amino-2-((3-carboxy-4-hydroxyphenyl)amino)benzoic Acid,N-(2-Carboxy-4-aminophenyl)-5-aminosalicylic Acid,2-Hydroxy-5-[(2-carboxy-4-aminophenyl)amino]benzoic Acid IUPAC Name: 5-amino-2-(3-carboxy-4-hydroxyanilino)benzoic acid SMILES: Nc1ccc(Nc2ccc(O)c(c2)C(=O)O)c(c1)C(=O)O