missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-[3-(trifluoromethyl)-3H-diazirin-3-yl]benzoic Acid, Methyl Ester, TRC

Product Code. 30394526
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394526 5 mg
30402842 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394526 Supplier Toronto Research Chemicals Supplier No. TRCH9715255MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165963-72-4
Molecular Formula C10 H7 F3 N2 O3
Molecular Weight (g/mol) 260.17
IUPAC Name methyl 2-hydroxy-4-[3-(trifluoromethyl)diazirin-3-yl]benzoate
SMILES COC(=O)c1ccc(cc1O)C2(N=N2)C(F)(F)F

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C10H7F3N2O3/c1-18-8(17)6-3-2-5(4-7(6)16)9(14-15-9)10(11,12)13/h2-4,16H,1H3
Formula Weight 260.0409
Chemical Name or Material 2-Hydroxy-4-[3-(trifluoromethyl)-3H-diazirin-3-yl]benzoic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.