CAS RN 165963-72-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Hydroxy-4-[3-(trifluoromethyl)-3H-diazirin-3-yl]benzoic Acid, Methyl Ester, TRC
CAS: 165963-72-4 Molecular Formula: C10 H7 F3 N2 O3 Molecular Weight (g/mol): 260.17 IUPAC Name: methyl 2-hydroxy-4-[3-(trifluoromethyl)diazirin-3-yl]benzoate SMILES: COC(=O)c1ccc(cc1O)C2(N=N2)C(F)(F)F