missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-methylcyclopent-2-enone, TRC

Product Code. 30392894
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392894 1 g -
30398330 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30392894 Supplier Toronto Research Chemicals Supplier No. TRCH2510401G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-71-7
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
Synonym 2-Cyclopenten-1-one, 2-hydroxy-3-methyl- (6CI, 7CI, 8CI, 9CI, ACI), 2-Hydroxy-3-methyl-2-cyclopenten-1-one (ACI), 2-Hydroxy-1-methyl-1-cyclopentene-3-one, 2-Hydroxy-1-methylcyclopenten-3-one, 2-Hydroxy-3-methyl-2-cyclopentene-1-one, 2-Hydroxy-3-methyl-2-cyclopentenone, 3-Methyl-2-hydroxy-2-cyclopenten-1-one, 3-Methyl-2-hydroxy-2-cyclopentenone, 3-Methylcyclopent-2-en-2-ol-1-one, Corylon, Corylone, Cycloten, Cyclotene, Cyclotene (odorant), Maple lactone, MCP, NSC 133445, NSC 84226
IUPAC Name 2-hydroxy-3-methylcyclopent-2-en-1-one
SMILES CC1=C(O)C(=O)CC1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H8O2/c1-4-2-3-5(7)6(4)8/h8H,2-3H2,1H3
Formula Weight 112.0524
Chemical Name or Material 2-Hydroxy-3-methylcyclopent-2-enone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.