accessibility menu, dialog, popup

UserName

CH3OHO

CAS RN 80-71-7

IUPAC Name: 2-hydroxy-3-methylcyclopent-2-en-1-one
Synonyms: methylcyclopentenolone
Molecular Weight (g/mol): 112.13
Molecular Formula: C6H8O2
InChi Key: CFAKWWQIUFSQFU-UHFFFAOYSA-N
SMILES: CC1=C(O)C(=O)CC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results