missing translation for 'onlineSavingsMsg'
Learn More

2-Hexadecyl-octadecanoic Acid Methyl Ester, TRC

Product Code. 30396562
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396562 250 mg -
30394076 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396562 Supplier Toronto Research Chemicals Supplier No. TRCH293805250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17658-53-6
Molecular Formula C35H70O2
Molecular Weight (g/mol) 522.93
Synonym 2-Hexadecyloctadecanoic Acid Methyl Ester, 2-Palmitylstearic Acid Methyl Ester
IUPAC Name methyl 2-hexadecyloctadecanoate
SMILES CCCCCCCCCCCCCCCCC(CCCCCCCCCCCCCCCC)C(=O)OC

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C35H70O2/c1-4-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34(35(36)37-3)33-31-29-27-25-23-21-19-17-15-13-11-9-7-5-2/h34H,4-33H2,1-3H3
Formula Weight 522.5376
Chemical Name or Material 2-Hexadecyl-octadecanoic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.