CAS RN 17658-53-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Hexadecyl-octadecanoic Acid Methyl Ester, TRC
CAS: 17658-53-6 Molecular Formula: C35H70O2 Molecular Weight (g/mol): 522.93 Synonym: 2-Hexadecyloctadecanoic Acid Methyl Ester,2-Palmitylstearic Acid Methyl Ester IUPAC Name: methyl 2-hexadecyloctadecanoate SMILES: CCCCCCCCCCCCCCCCC(CCCCCCCCCCCCCCCC)C(=O)OC