missing translation for 'onlineSavingsMsg'
Learn More

2-Formylphenylboronic acid pinacol ester, 97%

Product Code. 10442498 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10442498 1 g Glass bottle 1g
10319747 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10442498 Supplier Thermo Scientific Acros Supplier No. 430670010
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
This item is not returnable. View return policy

CAS: 380151-85-9 | C13H17BO3 | 232.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 380151-85-9
Molecular Formula C13H17BO3
Molecular Weight (g/mol) 232.09
MDL Number MFCD07363841
InChI Key SLJMPQLPRHIAOM-UHFFFAOYSA-N
Synonym 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 2-formylphenylboronic acid pinacol ester, 2-formylphenyl boronic acid pinacol ester, 2-formylphenylboronic acid, pinacol ester, 2-formylbenzeneboronic acid pinacol ester, 2-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 2-2-formylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzaldehyde, amtb063, 2-boronobenzaldehyde, pinacol ester
PubChem CID 10977322
IUPAC Name 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
SMILES CC1(C)OB(OC1(C)C)C1=CC=CC=C1C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 232.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Formylphenylboronic acid pinacol ester
Show More Show Less
compliance-icons
Product Identifier
  • 2-Formylphenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.