accessibility menu, dialog, popup

UserName

CH3H3COBOH3CH3CO

CAS RN 380151-85-9

IUPAC Name: 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Synonyms: 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Molecular Weight (g/mol): 232.09
Molecular Formula: C13H17BO3
InChi Key: SLJMPQLPRHIAOM-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=CC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results