missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorocinnamic acid, 98%, Thermo Scientific™

Product Code. 10300554 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10300554 1 g Glass bottle 1g
10606792 5 g Glass bottle 5g
10401831 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10300554 Supplier Thermo Scientific Acros Supplier No. 207100010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 451-69-4
Molecular Formula C9H7FO2
Molecular Weight (g/mol) 166.15
MDL Number MFCD00004370
InChI Key IOUDZAFBPDDAMK-AATRIKPKSA-N
Synonym 2-fluorocinnamic acid, o-fluorocinnamic acid, cinnamic acid, o-fluoro, 2-propenoic acid, 3-2-fluorophenyl, 2e-3-2-fluorophenyl prop-2-enoic acid, 3-2-fluorophenyl acrylic acid, 3-2-fluoro-phenyl-acrylic acid, 2e-3-2-fluorophenyl acrylic acid, 3-2-fluorophenyl-2-propenoic acid, trans-2-fluorocinnamic acid
PubChem CID 735833
IUPAC Name (E)-3-(2-fluorophenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=CC=CC=C1F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 179°C to 183°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula FC6H4CH=CHCO2H
Quantity 1 g
Beilstein 09, 594
Formula Weight 166.15
Percent Purity 98%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 2-Fluorocinnamic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluorocinnamic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.