Learn More
2-Fluorobiphenyl, 97%, Thermo Scientific™
£13.00 - £38.20
Chemical Identifiers
CAS | 321-60-8 |
---|---|
Molecular Formula | C12H9F |
Molecular Weight (g/mol) | 172.20 |
MDL Number | MFCD00000317 |
InChI Key | KLECYOQFQXJYBC-UHFFFAOYSA-N |
Synonym | 2-fluorobiphenyl, 2-fluoro-1,1'-biphenyl, o-fluorodiphenyl, 1,1'-biphenyl, 2-fluoro, biphenyl, 2-fluoro, ortho-fluorodiphenyl, 2-fluorodiphenyl, 2-fluoro-biphenyl, ccris 1659, 2-fluoro-1-phenylbenzene |
PubChem CID | 67579 |
SMILES | FC1=CC=CC=C1C1=CC=CC=C1 |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
321-60-8 | |
172.20 | |
KLECYOQFQXJYBC-UHFFFAOYSA-N | |
67579 |
C12H9F | |
MFCD00000317 | |
2-fluorobiphenyl, 2-fluoro-1,1'-biphenyl, o-fluorodiphenyl, 1,1'-biphenyl, 2-fluoro, biphenyl, 2-fluoro, ortho-fluorodiphenyl, 2-fluorodiphenyl, 2-fluoro-biphenyl, ccris 1659, 2-fluoro-1-phenylbenzene | |
FC1=CC=CC=C1C1=CC=CC=C1 |
Specifications
321-60-8 | |
100.0 | |
248.0°C | |
C12H9F | |
MFCD00000317 | |
2-fluorobiphenyl, 2-fluoro-1,1'-biphenyl, o-fluorodiphenyl, 1,1'-biphenyl, 2-fluoro, biphenyl, 2-fluoro, ortho-fluorodiphenyl, 2-fluorodiphenyl, 2-fluoro-biphenyl, ccris 1659, 2-fluoro-1-phenylbenzene | |
FC1=CC=CC=C1C1=CC=CC=C1 | |
172.20 | |
172.2 | |
2-Fluorobiphenyl, 97% |
96.0 | |
74.0°C to 76.0°C | |
97% | |
C6H5C6H4F | |
05,II,482 | |
KLECYOQFQXJYBC-UHFFFAOYSA-N | |
2-fluoro-1,1'-biphenyl | |
67579 | |
97% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN: Wash with plenty of soap and water. If skin irritation occurs: Get medical advice/attention. IF IN EYES: Rinse cautiously with water for several minutes. Remove con
missing translation for 'einecsNumber' : 206-290-7
missing translation for 'rtecsNumber' : DV5291000
missing translation for 'tsca' : TSCA
- 2-Fluorobiphenyl
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.