missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobiphenyl, 97%, Thermo Scientific™

Product Code. 10592138 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10592138 1 g Glass bottle 1g
10769438 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10592138 Supplier Thermo Scientific Acros Supplier No. 146840010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 321-60-8
Molecular Formula C12H9F
Molecular Weight (g/mol) 172.20
MDL Number MFCD00000317
InChI Key KLECYOQFQXJYBC-UHFFFAOYSA-N
Synonym 2-fluorobiphenyl, 2-fluoro-1,1'-biphenyl, o-fluorodiphenyl, 1,1'-biphenyl, 2-fluoro, biphenyl, 2-fluoro, ortho-fluorodiphenyl, 2-fluorodiphenyl, 2-fluoro-biphenyl, ccris 1659, 2-fluoro-1-phenylbenzene
PubChem CID 67579
SMILES FC1=CC=CC=C1C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 74.0°C to 76.0°C
Boiling Point 248.0°C
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5C6H4F
Quantity 1 g
Beilstein 05,II,482
IUPAC Name 2-fluoro-1,1'-biphenyl
Formula Weight 172.2
Percent Purity 97%
Chemical Name or Material 2-Fluorobiphenyl, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluorobiphenyl
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.