missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-m-xylene, 98%

Product Code. 11478623
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478623 25 g 25g
11468623 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478623 Supplier Thermo Scientific Alfa Aesar Supplier No. A10424.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 443-88-9 | C8H9F | 124.158 g/mol

It is used in the spectroscopic observation of jet-cooled 2-fluoro-m-xylyl radical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the spectroscopic observation of jet-cooled 2-fluoro-m-xylyl radical.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges and keep away from ignition sources.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 443-88-9
Molecular Formula C8H9F
Molecular Weight (g/mol) 124.158
MDL Number MFCD00039215
InChI Key JTAUTNBVFDTYTI-UHFFFAOYSA-N
Synonym 2,6-dimethylfluorobenzene, 2-fluoro-m-xylene, 1-fluoro-2,6-dimethylbenzene, 1,3-dimethyl-2-fluorobenzene, benzene, 2-fluoro-1,3-dimethyl, 2,6-dimethyl fluorobenzene, 2-fluoro-1,3-dimethyl-benzene, fr b1 f1, pubchem4407, 2-fluoro-3-methyltoluene
PubChem CID 123065
IUPAC Name 2-fluoro-1,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)C)F

Specifications

Density 0.988
Quantity 25 g
Odor Aromatic
Refractive Index 1.479
UN Number UN1993
Beilstein 1857413
Boiling Point 147°C to 148°C
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 124.16
Percent Purity 98%
Chemical Name or Material 2-Fluoro-m-xylene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.