CAS RN 443-88-9
IUPAC Name:
2-fluoro-1,3-dimethylbenzene
Synonyms:
2-fluoro-1,3-dimethylbenzene
Molecular Weight (g/mol):
124.16
Molecular Formula:
C8H9F
InChi Key:
JTAUTNBVFDTYTI-UHFFFAOYSA-N
SMILES:
CC1=CC=CC(C)=C1F
1
–
2
of
2
results
1
2-Fluoro-m-xylene, 98%
CAS: 443-88-9 Molecular Formula: C8H9F Molecular Weight (g/mol): 124.158 MDL Number: MFCD00039215 InChI Key: JTAUTNBVFDTYTI-UHFFFAOYSA-N Synonym: 2,6-dimethylfluorobenzene,2-fluoro-m-xylene,1-fluoro-2,6-dimethylbenzene,1,3-dimethyl-2-fluorobenzene,benzene, 2-fluoro-1,3-dimethyl,2,6-dimethyl fluorobenzene,2-fluoro-1,3-dimethyl-benzene,fr b1 f1,pubchem4407,2-fluoro-3-methyltoluene PubChem CID: 123065 IUPAC Name: 2-fluoro-1,3-dimethylbenzene SMILES: CC1=C(C(=CC=C1)C)F