missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-DL-phenylalanine, 98%

Product Code. 11342007
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342007 5 g 5g
11332007 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342007 Supplier Thermo Scientific Alfa Aesar Supplier No. L07654.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2629-55-2 | C9H10FNO2 | 183.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2629-55-2
Molecular Formula C9H10FNO2
Molecular Weight (g/mol) 183.182
MDL Number MFCD00004271
InChI Key NYCRCTMDYITATC-UHFFFAOYSA-N
Synonym 2-fluoro-dl-phenylalanine, 2-amino-3-2-fluorophenyl propanoic acid, 2-fluorophenylalanine, o-fluoro-dl-phenylalanine, o-fluorophenylalanine, 3-o-fluorophenyl alanine, dl-2-fluorophenyl alanine, dl-2-f-phe-oh, dl-o-fluorophenylalanine, dl-beta-o-fluorophenylalanine
PubChem CID 9465
IUPAC Name 2-amino-3-(2-fluorophenyl)propanoic acid
SMILES C1=CC=C(C(=C1)CC(C(=O)O)N)F

Specifications

Melting Point ∽245°C (decomposition)
Beilstein 3201186
Quantity 5 g
Formula Weight 183.18
Percent Purity 98%
Chemical Name or Material 2-Fluoro-DL-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.