accessibility menu, dialog, popup

UserName

CAS RN 2629-55-2

IUPAC Name: (2S)-2-azaniumyl-3-(2-fluorophenyl)propanoate
Synonyms: (2S)-2-ammonio-3-(2-fluorophenyl)propanoate
Molecular Weight (g/mol): 183.18
Molecular Formula: C9H10FNO2
InChi Key: NYCRCTMDYITATC-QMMMGPOBSA-N
SMILES: [NH3+][C@@H](CC1=CC=CC=C1F)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results