missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Fluoro-2'-deoxyguanosine, 99%

Product Code. 15446179
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15446179 250 mg 250mg
15436179 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15446179 Supplier Thermo Scientific Chemicals Supplier No. J64055.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 78842-13-4 | C10H12FN5O4 | 285.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78842-13-4
Molecular Formula C10H12FN5O4
Molecular Weight (g/mol) 285.24
MDL Number MFCD00923832
InChI Key UXUZARPLRQRNNX-SJLGBTOWNA-N
Synonym 2'-deoxy-2'-fluoroguanosine, 2'-fluoro-2'-deoxyguanosine, guanosine, 2'-deoxy-2'-fluoro, 2-amino-9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl-tetrahydro-furan-2-yl-1,9-dihydro-purin-6-one, 2-amino-9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-purin-6-one, 2-amino-9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one, 2'-dfgs, deoxyfluoroguanosine, 2-amino-9-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-1h-purin-6-one
PubChem CID 196536
IUPAC Name 2-amino-9-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES NC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2F)C(=O)N1

Specifications

Color White
Quantity 250 mg
Formula Weight 285.23
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2'-Fluoro-2'-deoxyguanosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.