missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoro-2'-deoxyadenosine, TRC

Product Code. 30407103
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Product Code. Quantity
30407103 500 mg
30397822 50 mg
30393602 100 mg
3 options
This item is not returnable. View return policy

Product Code. 30407103

Brand: Toronto Research Chemicals TRCF590805500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64183-27-3
Formula Weight 269.09
InChI Formula InChI=1S/C10H12FN5O3/c11-5-7(18)4(1-17)19-10(5)16-3-15-6-8(12)13-2-14-9(6)16/h2-5,7,10,17-18H,1H2,(H2,12,13,14)/t4-,5-,7-,10-/m1/s1
IUPAC Name (2R,3R,4R,5R)-5-(6-amino-9H-purin-9-yl)-4-fluoro-2-(hydroxymethyl)tetrahydrofuran-3-ol
Molecular Formula C10H12FN5O3
Molecular Weight (g/mol) 269.24
SMILES F[C@H]1[C@H](N2C=3C(N=C2)=C(N)N=CN3)O[C@H](CO)[C@H]1O
Synonym 2-Fluoro-2-Deoxyadenosine
Chemical Name or Material 2'-Fluoro-2'-deoxyadenosine

Specifications

Quantity 500 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.