Learn More
2-Ethylhexanal, 96%, Thermo Scientific Chemicals
£49.50 - £139.00
Chemical Identifiers
CAS | 123-05-7 |
---|---|
Molecular Formula | C8H16O |
Molecular Weight (g/mol) | 128.21 |
MDL Number | MFCD00006987 |
InChI Key | LGYNIFWIKSEESD-UHFFFAOYSA-N |
Synonym | hexanal, 2-ethyl, 2-ethylhexaldehyde, 2-ethylhexylaldehyde, 3-formylheptane, 2-ethylcaproaldehyde, butylethylacetaldehyde, ethylbutylacetaldehyde, ethylhexaldehyde, alpha-ethylcaproaldehyde, butyl ethyl acetaldehyde |
PubChem CID | 31241 |
IUPAC Name | 2-ethylhexanal |
SMILES | CCCCC(CC)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10269790
|
Thermo Scientific Acros
410092500 |
250 g | Glass bottle |
£49.50
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10635491
|
Thermo Scientific Acros
410090010 |
1 kg | Glass bottle |
£139.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
123-05-7 | |
128.21 | |
LGYNIFWIKSEESD-UHFFFAOYSA-N | |
31241 | |
CCCCC(CC)C=O |
C8H16O | |
MFCD00006987 | |
hexanal, 2-ethyl, 2-ethylhexaldehyde, 2-ethylhexylaldehyde, 3-formylheptane, 2-ethylcaproaldehyde, butylethylacetaldehyde, ethylbutylacetaldehyde, ethylhexaldehyde, alpha-ethylcaproaldehyde, butyl ethyl acetaldehyde | |
2-ethylhexanal |
Specifications
8mg KOH/g max. | |
95.0 | |
-90°C | |
0.8195g/mL | |
50°C | |
95% min. (GC) | |
CH3(CH2)3CH(C2H5)CHO | |
MFCD00006987 | |
hexanal, 2-ethyl, 2-ethylhexaldehyde, 2-ethylhexylaldehyde, 3-formylheptane, 2-ethylcaproaldehyde, butylethylacetaldehyde, ethylbutylacetaldehyde, ethylhexaldehyde, alpha-ethylcaproaldehyde, butyl ethyl acetaldehyde | |
LGYNIFWIKSEESD-UHFFFAOYSA-N | |
2-ethylhexanal | |
31241 | |
96% | |
2-Ethylhexanal, pract., 99% |
123-05-7 | |
100.0 | |
Colorless | |
162.0°C to 165.0°C | |
Authentic | |
C8H16O | |
1.4145 to 1.4175 | |
0.8195 | |
Solubility in water: < 0.2g/L (20°C). Other solubilities: misccible with most organic solvents | |
CCCCC(CC)C=O | |
128.21 | |
128.21 | |
Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
May cause an allergic skin reaction.
Suspected of damaging fertility or the unborn child.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Obtain special instructions before use.
Wea
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 204-596-5
missing translation for 'rtecsNumber' : MN7525000
missing translation for 'tsca' : TSCA
- 2-Ethylhexanal
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only