missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexanal, 96%

Product Code. 10269790 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10269790 250 g Glass bottle 250g
10635491 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10269790 Supplier Thermo Scientific Acros Supplier No. 410092500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 123-05-7 | C8H16O | 128.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-05-7
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.21
MDL Number MFCD00006987
InChI Key LGYNIFWIKSEESD-UHFFFAOYSA-N
Synonym hexanal, 2-ethyl, 2-ethylhexaldehyde, 2-ethylhexylaldehyde, 3-formylheptane, 2-ethylcaproaldehyde, butylethylacetaldehyde, ethylbutylacetaldehyde, ethylhexaldehyde, alpha-ethylcaproaldehyde, butyl ethyl acetaldehyde
PubChem CID 31241
IUPAC Name 2-ethylhexanal
SMILES CCCCC(CC)C=O

Specifications

Acidity 8mg KOH/g max.
CAS Min % 95.0
CAS Max % 100.0
Melting Point -90°C
Color Colorless
Density 0.8195g/mL
Boiling Point 162.0°C to 165.0°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3CH(C2H5)CHO
Refractive Index 1.4145 to 1.4175
Quantity 250 g
Specific Gravity 0.8195
Solubility Information Solubility in water: < 0.2g/L (20°C). Other solubilities: misccible with most organic solvents
Formula Weight 128.21
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2-Ethylhexanal, pract., 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Ethylhexanal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.