missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxypropene, TRC

Product Code. 30376326
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30376326 25 g
30385086 100 g
30371237 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30376326 Supplier Toronto Research Chemicals Supplier No. TRCE89264025G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 926-66-9
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
Synonym 2-Ethoxy-1-propene, 2-Ethoxypropene, Ethyl 1-Methylvinyl Ether, Ethyl Isopropenyl Ether
SMILES CCOC(=C)C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C5H10O/c1-4-6-5(2)3/h2,4H2,1,3H3
Formula Weight 86.0732
Chemical Name or Material 2-Ethoxypropene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.