CAS RN 926-66-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Ethoxypropene, TRC
CAS: 926-66-9 Molecular Formula: C5H10O Molecular Weight (g/mol): 86.13 Synonym: 2-Ethoxy-1-propene,2-Ethoxypropene,Ethyl 1-Methylvinyl Ether,Ethyl Isopropenyl Ether SMILES: CCOC(=C)C