missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxybenzyl alcohol, 98%

Product Code. 11452847
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452847 25 g 25g
11462847 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11452847 Supplier Thermo Scientific Alfa Aesar Supplier No. A15477.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 71672-75-8 | C9H12O2 | 152.19 g/mol

2-Ethoxybenzyl alcohol undergoes acetylation with acetic anhydride over zeolite to yield O-acetylated product. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethoxybenzyl alcohol undergoes acetylation with acetic anhydride over zeolite to yield O-acetylated product. It is also used as pharmaceutical intermediates.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71672-75-8
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00004616
InChI Key ICJVQAHPHKYCNU-UHFFFAOYSA-N
Synonym 2-ethoxyphenyl methanol, 2-ethoxybenzyl alcohol, benzenemethanol, 2-ethoxy, o-ethoxybenzyl alcohol, ethyl ether of saligenin, 2-ethoxybenzylalcohol, 2-ethoxybenzenemethanol, benzyl alcohol, o-ethoxy, benzenemethanol, 2-ethoxy-9ci, acmc-1bgrn
PubChem CID 51381
IUPAC Name (2-ethoxyphenyl)methanol
SMILES CCOC1=CC=CC=C1CO

Specifications

Density 1.074
Boiling Point 263°C to 265°C
Flash Point >110°C (230°F)
Refractive Index 1.532
Quantity 25 g
Beilstein 2555374
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 2-Ethoxybenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.