CAS RN 71672-75-8
CAS RN 71672-75-8
IUPAC Name:
(2-ethoxyphenyl)methanol
Synonyms:
(2-ethoxyphenyl)methanol
Molecular Weight (g/mol):
152.19
Molecular Formula:
C9H12O2
InChi Key:
ICJVQAHPHKYCNU-UHFFFAOYSA-N
SMILES:
CCOC1=CC=CC=C1CO
1
–
1
of
1
results
1
2-Ethoxybenzyl alcohol, 98%
CAS: 71672-75-8 Molecular Formula: C9H12O2 Molecular Weight (g/mol): 152.19 MDL Number: MFCD00004616 InChI Key: ICJVQAHPHKYCNU-UHFFFAOYSA-N Synonym: 2-ethoxyphenyl methanol,2-ethoxybenzyl alcohol,benzenemethanol, 2-ethoxy,o-ethoxybenzyl alcohol,ethyl ether of saligenin,2-ethoxybenzylalcohol,2-ethoxybenzenemethanol,benzyl alcohol, o-ethoxy,benzenemethanol, 2-ethoxy-9ci,acmc-1bgrn PubChem CID: 51381 IUPAC Name: (2-ethoxyphenyl)methanol SMILES: CCOC1=CC=CC=C1CO