missing translation for 'onlineSavingsMsg'
Learn More

2-Dodecyltetradecanoic Acid, TRC

Product Code. 30383702
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383702 2.5 g 2.5g
30377761 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383702 Supplier Toronto Research Chemicals Supplier No. TRCD4417752.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24251-98-7
Molecular Formula C26H52O2
Molecular Weight (g/mol) 396.69
IUPAC Name 2-dodecyltetradecanoic acid
SMILES CCCCCCCCCCCCC(CCCCCCCCCCCC)C(=O)O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C26H52O2/c1-3-5-7-9-11-13-15-17-19-21-23-25(26(27)28)24-22-20-18-16-14-12-10-8-6-4-2/h25H,3-24H2,1-2H3,(H,27,28)
Formula Weight 396.3967
Chemical Name or Material 2-Dodecyltetradecanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.