CAS RN 24251-98-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Dodecyltetradecanoic Acid, TRC
CAS: 24251-98-7 Molecular Formula: C26H52O2 Molecular Weight (g/mol): 396.69 IUPAC Name: 2-dodecyltetradecanoic acid SMILES: CCCCCCCCCCCCC(CCCCCCCCCCCC)C(=O)O