missing translation for 'onlineSavingsMsg'
Learn More

2-(Diphenylphosphino)ethyltriethoxysilane, 92%

Product Code. 10435085 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10435085 1 g Glass bottle 1g
10443745 5 g Glass bottle 5g
10686494 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10435085 Supplier Thermo Scientific Acros Supplier No. 391860010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 18586-39-5 | C20H29O3PSi | 376.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18586-39-5
Molecular Formula C20H29O3PSi
Molecular Weight (g/mol) 376.51
MDL Number MFCD00015125
InChI Key HLXCYTXLQJWQFG-UHFFFAOYSA-N
Synonym 2-diphenylphosphino ethyltriethoxysilane, diphenyl 2-triethoxysilyl ethyl phosphine, phosphine, diphenyl 2-triethoxysilyl ethyl, diphenyl 2-triethoxysilyl ethyl phosphane, phosphine,diphenyl 2-triethoxysilyl ethyl, triethoxysilyl ethyl diphenylphosphine, ksc268s0d
PubChem CID 87708
IUPAC Name diphenyl(2-triethoxysilylethyl)phosphane
SMILES CCO[Si](CCP(C1=CC=CC=C1)C1=CC=CC=C1)(OCC)OCC

Specifications

CAS Min % 90.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 182.0°C (1.3 mmHg)
Flash Point 134.5°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Refractive Index 1.5390 to 1.5410 (20°C, 589nm)
Quantity 1 g
Specific Gravity 1.05
Solubility Information Solubility in water: reacts.
Formula Weight 376.51
Percent Purity 92%
Physical Form Liquid
Chemical Name or Material 2-(Diphenylphosphino)ethyltriethoxysilane
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Diphenylphosphino)ethyltriethoxysilane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.