accessibility menu, dialog, popup

UserName

CAS RN 18586-39-5

IUPAC Name: diphenyl[2-(triethoxysilyl)ethyl]phosphane
Synonyms: diphenyl[2-(triethoxysilyl)ethyl]phosphane
Molecular Weight (g/mol): 376.51
Molecular Formula: C20H29O3PSi
InChi Key: HLXCYTXLQJWQFG-UHFFFAOYSA-N
SMILES: CCO[Si](CCP(C1=CC=CC=C1)C1=CC=CC=C1)(OCC)OCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results