missing translation for 'onlineSavingsMsg'
Learn More

2-(Diphenylphosphino)ethyltriethoxysilane, 92%, Thermo Scientific Chemicals

Product Code. 10443745
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18586-39-5
Molecular Formula C20H29O3PSi
Molecular Weight (g/mol) 376.51
MDL Number MFCD00015125
InChI Key HLXCYTXLQJWQFG-UHFFFAOYSA-N
Synonym 2-diphenylphosphino ethyltriethoxysilane, diphenyl 2-triethoxysilyl ethyl phosphine, phosphine, diphenyl 2-triethoxysilyl ethyl, diphenyl 2-triethoxysilyl ethyl phosphane, phosphine,diphenyl 2-triethoxysilyl ethyl, triethoxysilyl ethyl diphenylphosphine, ksc268s0d
PubChem CID 87708
IUPAC Name diphenyl(2-triethoxysilylethyl)phosphane
SMILES CCO[Si](CCP(C1=CC=CC=C1)C1=CC=CC=C1)(OCC)OCC

Specifications

CAS Min % 90.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 182.0°C (1.3 mmHg)
Flash Point 134.5°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Refractive Index 1.5390 to 1.5410 (20°C, 589nm)
Quantity 5 g
Specific Gravity 1.05
Solubility Information Solubility in water: reacts.
Formula Weight 376.51
Percent Purity 92%
Physical Form Liquid
Chemical Name or Material 2-(Diphenylphosphino)ethyltriethoxysilane
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Diphenylphosphino)ethyltriethoxysilane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'