missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxyinosine, 98+%

Product Code. 15438917
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438917 250 mg 250mg
15418917 1 g 1g
15428917 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15438917 Supplier Thermo Scientific Chemicals Supplier No. H52292.MD
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 890-38-0 | C10H12N4O4 | 252.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 890-38-0
Molecular Formula C10H12N4O4
Molecular Weight (g/mol) 252.23
MDL Number MFCD00005762
InChI Key VGONTNSXDCQUGY-YUZWJPFSNA-N
Synonym 2'-deoxyinosine, deoxyinosine, inosine, 2'-deoxy, 2-deoxyinosine, 9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-ol, unii-hn0rq6sbwq, delta-ino, d-ino, hn0rq6sbwq, 9-2-deoxy-beta-d-ribofuranosyl hypoxanthine
PubChem CID 65058
ChEBI CHEBI:28997
IUPAC Name 9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C1N=CNC2=O

Specifications

Quantity 250 mg
Formula Weight 252.23
Percent Purity ≥98%
Chemical Name or Material 2'-Deoxyinosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.