missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyguanosine Monohydrate, Synthetic, >98% (Dry Basis)

Product Code. 15825458 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15825458 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15825458 Supplier Thermo Scientific Supplier No. J14304.09
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 312693-72-4 | C10H15N5O5 | 285.26 g/mol

 

TRUSTED_SUSTAINABILITY

Specifications

CAS 312693-72-4
Packaging Plastic bottle
Molecular Formula C10H15N5O5
MDL Number MFCD00150760
Quantity 10 g
Synonym 2 '-deoxyguanosine monohydrate, 2 '-deoxyguanosine hydrate, 2-deoxyguanosine hydrate, 2-amino-9-2 r,4 s,5 r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1 h-purin-6 9 h-one hydrate, 2-amino-9-2 r,4 s,5 r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1 h-purin-6 9 h-one hydrate 1:x, 2 acute-deoxyguanosine, deoxyguanosine hydrate, 2'deoxyguanosine monohydrate, 2'-deoxyguanosinemonohydrate
InChI Key LZSCQUCOIRGCEJ-VDPVQVSMNA-N
SMILES O.NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1
IUPAC Name 2-amino-9-[(2 R,4 S,5 R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3 H-purin-6-one;hydrate
Molecular Weight (g/mol) 285.26
PubChem CID 16218597
Formula Weight 285.26
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material 2 '-Deoxyguanosine monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.