missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyadenosine Monohydrate-13C5, TRC

Product Code. 30133612
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133612 1 mg 1mg
30009937 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133612 Supplier Toronto Research Chemicals Supplier No. TRCD2316221MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 478510-79-1
Molecular Formula C513C5H13N5O3•H2O
Molecular Weight (g/mol) 274.22
Synonym 2’-Deoxy-β-D-adenosine-13C5 Monohydrate, 9-(2-Deoxy-β-D-erythro-pentofuranosyl) adenine-13C5, 9-(2-Deoxy-β-D-erythro-pentofuranosyl)-9H-purin-6-amine-13C5 Hydrate, Adenine Deoxyribonucleoside-13C5 Hydrate, Adenine Deoxyribose-13C5 Hydrate, Adenyldeoxyriboside-13C5 Hydrate, Deoxyadenosine-13C5 Hydrate, Desoxyadenosine-13C5 Hydrate, NSC 141848, NSC 143510, NSC 83258, dA, 1-(6-Amino-9H-purin-9-yl)-1,2-dideoxy-β-D-ribofuranose-13C5 Hydrate, 2-Deoxyadenine-9-β-D-erythro-pento-furanoside-13C5 Hydrate;
SMILES NC1=NC=NC2=C1N=CN2[13C@]3[13C][13C@](O)[13C@@]([13C]O)O3.O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C10H13N5O3.H2O/c11-9-8-10(13-3-12-9)15(4-14-8)7-1-5(17)6(2-16)18-7;/h3-7,16-17H,1-2H2,(H2,11,12,13);1H2
Percent Purity >95%
Chemical Name or Material 2'-Deoxyadenosine Monohydrate-13C5
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.