missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopentene-1-acetic acid, tech. 90%, remainder mainly 3-cyclopentene-1-acetic acid

Product Code. 11431760
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431760 5 g 5g
11441760 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11431760 Supplier Thermo Scientific Alfa Aesar Supplier No. H27507.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 13668-61-6 | C7H10O2 | 126.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13668-61-6
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00001400
InChI Key NNRZTJAACCRFRV-UHFFFAOYSA-N
Synonym 2-cyclopentene-1-acetic acid, 2-cyclopent-2-en-1-yl acetic acid, cyclopent-2-enylacetic acid, 2-cyclopenten-1-ylacetic acid, cyclopent-2-en-1-ylacetic acid, cyclopentene-3-acetic acid, 2-cyclopentene-1-aceticacid, acmc-1c47k, 2-cyclopent-2-enylacetic acid, cyclopent-2-ene-1-acetic acid
PubChem CID 96216
IUPAC Name 2-cyclopent-2-en-1-ylacetic acid
SMILES C1CC(C=C1)CC(=O)O

Specifications

Melting Point 19°C
Density 1.047
Boiling Point 238°C
Flash Point 143°C (289°F)
Odor Rancid
Refractive Index 1.468
Quantity 5 g
Beilstein 2042172
Formula Weight 126.15
Percent Purity 90%
Grade Technical
Assay remainder mainly 3-cyclopentene-1-acetic acid
Chemical Name or Material 2-Cyclopentene-1-acetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.