accessibility menu, dialog, popup

UserName

CAS RN 13668-61-6

OO(S)

CAS RN 13668-61-6

IUPAC Name: 2-[(1S)-cyclopent-2-en-1-yl]acetate
Synonyms: 2-[(1S)-cyclopent-2-en-1-yl]acetate
Molecular Weight (g/mol): 125.15
Molecular Formula: C7H9O2
InChi Key: NNRZTJAACCRFRV-LURJTMIESA-M
SMILES: [O-]C(=O)C[C@@H]1CCC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results