missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methylpyridine-3-boronic acid, 98%, Thermo Scientific™

Product Code. 15575176 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15575176

Brand: Thermo Scientific Acros 455940010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 913835-86-6
Molecular Formula C6H7BClNO2
Molecular Weight (g/mol) 171.39
InChI Key ZMHWEBDZKPQNHG-UHFFFAOYSA-N
Synonym 2-chloro-5-methylpyridine-3-boronic acid, 2-chloro-5-methylpyridin-3-yl boronic acid, 2-chloro-5-picoline-3-boronic acid, 2-chloro-5-methylpyridine-3-boronicacid, alphachiron 13000b651, boronicacid, b-2-chloro-5-methyl-3-pyridinyl, 2-chloro-5-methyl-3-pyridineboronic acid, pubchem11116, acmc-209ra8, abbypharma ap-16-5787
PubChem CID 2763126
IUPAC Name (2-chloro-5-methylpyridin-3-yl)boronic acid
SMILES B(C1=CC(=CN=C1Cl)C)(O)O

Specifications

Melting Point 115.5°C to 119.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 171.39
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-methylpyridine-3-boronic acid
compliance-icons
Product Identifier
  • 2-Chloro-5-methylpyridine-3-boronic acid, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.