accessibility menu, dialog, popup

UserName

CAS RN 913835-86-6

BNClCH3HOOH

CAS RN 913835-86-6

IUPAC Name: (2-chloro-5-methylpyridin-3-yl)boronic acid
Synonyms: 2-chloro-5-methylpyridin-3-ylboronic acid
Molecular Weight (g/mol): 171.39
Molecular Formula: C6H7BClNO2
InChi Key: ZMHWEBDZKPQNHG-UHFFFAOYSA-N
SMILES: CC1=CN=C(Cl)C(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results