missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluorophenylacetic acid, 97%, Thermo Scientific™

Product Code. 11396909
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11396909

Brand: Thermo Scientific Alfa Aesar H26803.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177985-33-0
Molecular Formula C8H6ClFO2
Molecular Weight (g/mol) 188.58
MDL Number MFCD01016155
InChI Key VFUKAVLGRHSSPG-UHFFFAOYSA-N
Synonym 2-chloro-5-fluorophenylacetic acid, 2-2-chloro-5-fluorophenyl acetic acid, 2-chloro-5-fluorophenyl acetic acid, pubchem2711, 2-2-chloro-5-fluoro-phenyl acetic acid, ksc497i1n, 2-chloro-5-fluoro-phenyl-acetic acid
PubChem CID 3535430
IUPAC Name 2-(2-chloro-5-fluorophenyl)acetic acid
SMILES OC(=O)CC1=CC(F)=CC=C1Cl

Specifications

Quantity 1 g
Formula Weight 188.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-fluorophenylacetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.