CAS RN 177985-33-0
CAS RN 177985-33-0
IUPAC Name:
2-(2-chloro-5-fluorophenyl)acetic acid
Synonyms:
(2-chloro-5-fluorophenyl)acetic acid
Molecular Weight (g/mol):
188.58
Molecular Formula:
C8H6ClFO2
InChi Key:
VFUKAVLGRHSSPG-UHFFFAOYSA-N
SMILES:
OC(=O)CC1=CC(F)=CC=C1Cl
1
–
1
of
1
results
1
2-Chloro-5-fluorophenylacetic acid, 97%, Thermo Scientific™
CAS: 177985-33-0 Molecular Formula: C8H6ClFO2 Molecular Weight (g/mol): 188.58 MDL Number: MFCD01016155 InChI Key: VFUKAVLGRHSSPG-UHFFFAOYSA-N Synonym: 2-chloro-5-fluorophenylacetic acid,2-2-chloro-5-fluorophenyl acetic acid,2-chloro-5-fluorophenyl acetic acid,pubchem2711,2-2-chloro-5-fluoro-phenyl acetic acid,ksc497i1n,2-chloro-5-fluoro-phenyl-acetic acid PubChem CID: 3535430 IUPAC Name: 2-(2-chloro-5-fluorophenyl)acetic acid SMILES: OC(=O)CC1=CC(F)=CC=C1Cl