missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3,6-difluorobenzoyl chloride, 97%, Thermo Scientific™

Product Code. 11324935
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324935 1 g 1g
11334935 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11324935 Supplier Thermo Scientific Alfa Aesar Supplier No. L18169.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261762-42-9
Molecular Formula C7H2Cl2F2O
Molecular Weight (g/mol) 210.989
MDL Number MFCD01631442
InChI Key CBFPZKKUVNEIFF-UHFFFAOYSA-N
Synonym 2-chloro-3,6-difluorobenzoylchloride, acmc-20ao08, 2-chloro-3,6,-difluorobenzoyl chloride, benzoyl chloride,2-chloro-3,6-difluoro, 2-chloranyl-3,6-bis fluoranyl benzoyl chloride, benzoyl chloride, 2-chloro-3,6-difluoro-9ci
PubChem CID 2773536
IUPAC Name 2-chloro-3,6-difluorobenzoyl chloride
SMILES C1=CC(=C(C(=C1F)C(=O)Cl)Cl)F

Specifications

Refractive Index 1.51
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 211
Percent Purity 97%
Chemical Name or Material 2-Chloro-3,6-difluorobenzoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.