accessibility menu, dialog, popup

UserName

CAS RN 261762-42-9

FFClOCl

CAS RN 261762-42-9

IUPAC Name: 2-chloro-3,6-difluorobenzoyl chloride
Synonyms: 2-chloro-3,6-difluorobenzoyl chloride
Molecular Weight (g/mol): 210.99
Molecular Formula: C7H2Cl2F2O
InChi Key: CBFPZKKUVNEIFF-UHFFFAOYSA-N
SMILES: FC1=CC=C(F)C(C(Cl)=O)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results