missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3,4-dimethoxybenzyl alcohol, 97%

Product Code. 11449157
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449157 1 g 1g
11455526 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449157 Supplier Thermo Scientific Alfa Aesar Supplier No. B20688.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 93983-13-2 | C9H11ClO3 | 202.63 g/mol

It is applied in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in pharmaceutical industry.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93983-13-2
Molecular Formula C9H11ClO3
Molecular Weight (g/mol) 202.63
MDL Number MFCD00210141
InChI Key ZXYBQLFOEYWYBM-UHFFFAOYSA-N
Synonym 2-chloro-3,4-dimethoxyphenyl methanol, 2-chloro-3,4-dimethoxybenzyl alcohol, 2-chloroveratryl alcohol, maybridge1_002248, 2-chloro-3,4-dimethoxybenzenemethanol, benzenemethanol,2-chloro-3,4-dimethoxy, 2-chloro-3,4-dimethoxyphenyl methan-1-ol, 2-chloro-3,4-bis methyloxy phenyl methanol
PubChem CID 2775134
IUPAC Name (2-chloro-3,4-dimethoxyphenyl)methanol
SMILES COC1=CC=C(CO)C(Cl)=C1OC

Specifications

Melting Point 70°C to 72°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 202.64
Percent Purity 97%
Chemical Name or Material 2-Chloro-3,4-dimethoxybenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.