accessibility menu, dialog, popup

UserName

H3COOHClOCH3

CAS RN 93983-13-2

IUPAC Name: (2-chloro-3,4-dimethoxyphenyl)methanol
Synonyms: (2-chloro-3,4-dimethoxyphenyl)methanol
Molecular Weight (g/mol): 202.63
Molecular Formula: C9H11ClO3
InChi Key: ZXYBQLFOEYWYBM-UHFFFAOYSA-N
SMILES: COC1=CC=C(CO)C(Cl)=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results