missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2'-arabino-fluoro-2'-deoxyadenosine, 99+%, Thermo Scientific Chemicals

Catalog Number 15470157
Click to view available options
Quantity:
50 mg
100 mg
250 mg
This item is not returnable. View return policy
123318-82-1
C10H12ClN5O3
285.69
MFCD00871077
ADXUXRNLBYKGOA-GPGUAWMRNA-N
clofarabine, clolar, evoltra, clofarex, cafda, clofarabina, clofarabinum, clofarabine usan, cl-f-ara-a, c1-f-ara-a
119182
CHEBI:681569
(2R,3R,4S,5R)-5-(6-amino-2-chloropurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol
NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3Cl)C2=NC=N1
This item is not returnable. View return policy
123318-82-1
C10H12ClN5O3
285.69
MFCD00871077
ADXUXRNLBYKGOA-GPGUAWMRNA-N
clofarabine, clolar, evoltra, clofarex, cafda, clofarabina, clofarabinum, clofarabine usan, cl-f-ara-a, c1-f-ara-a
119182
CHEBI:681569
(2R,3R,4S,5R)-5-(6-amino-2-chloropurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol
NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3Cl)C2=NC=N1

A purine nucleoside anti-metabolite. Toxic to nondividing lymphocytes and monocytes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A purine nucleoside anti-metabolite
  • Toxic to nondividing lymphocytes and monocytes
TRUSTED_SUSTAINABILITY
Color White
Quantity 50 mg
UN Number UN2811
Merck Index 14,2372
Formula Weight 303.68
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material 2-Chloro-2'-arabino-fluoro-2'-deoxyadenosine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'