missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2'-arabino-fluoro-2'-deoxyadenosine, 99+%, Thermo Scientific Chemicals

Product Code. 15460157
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
50 mg
100 mg
250 mg
missing translation for 'unitSize':
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
15460157 250 mg 250mg
15470157 50 mg 50mg
15450157 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 15460157 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' J60927.MD
 Restricted Item
Request Bulk or Custom Formats

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 123318-82-1 | Molecular Formula: C10H12ClN5O3 | Molecular Weight: 285.69 g/mol

A purine nucleoside anti-metabolite. Toxic to nondividing lymphocytes and monocytes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A purine nucleoside anti-metabolite
  • Toxic to nondividing lymphocytes and monocytes
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123318-82-1
Molecular Formula C10H12ClN5O3
Molecular Weight (g/mol) 285.69
MDL Number MFCD00871077
InChI Key ADXUXRNLBYKGOA-GPGUAWMRNA-N
Synonym clofarabine, clolar, evoltra, clofarex, cafda, clofarabina, clofarabinum, clofarabine usan, cl-f-ara-a, c1-f-ara-a
PubChem CID 119182
ChEBI CHEBI:681569
IUPAC Name (2R,3R,4S,5R)-5-(6-amino-2-chloropurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3Cl)C2=NC=N1

Specifications

Color White
Quantity 250 mg
UN Number UN2811
Merck Index 14,2372
Formula Weight 303.68
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material 2-Chloro-2'-arabino-fluoro-2'-deoxyadenosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.