missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2,2-difluoroacetamide, 98%

Product Code. 11340214
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11340214 5 g 5g
11350214 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11340214 Supplier Thermo Scientific Alfa Aesar Supplier No. L00785.06
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 354-28-9 | C2H2ClF2NO | 129.491 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 354-28-9
Molecular Formula C2H2ClF2NO
Molecular Weight (g/mol) 129.491
MDL Number MFCD00039775
InChI Key PSAKKOKLSDIKEK-UHFFFAOYSA-N
Synonym chlorodifluoroacetamide, acetamide, 2-chloro-2,2-difluoro, difluorochloroacetamide, snylgqdjptadtaetp@, acmc-20algv, acetamide,2-difluoro, pubchem14583, 2-chloranyl-2,2-bis fluoranyl ethanamide
PubChem CID 67715
IUPAC Name 2-chloro-2,2-difluoroacetamide
SMILES C(=O)(C(F)(F)Cl)N

Specifications

Melting Point 79°C to 84°C
Quantity 5 g
Beilstein 1753808
Formula Weight 129.49
Percent Purity 98%
Chemical Name or Material 2-Chloro-2,2-difluoroacetamide
compliance-icons
Product Identifier
  • 2-Chloro-2,2-difluoroacetamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-Chloro-2,2-difluoroacetamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.