accessibility menu, dialog, popup

UserName

CAS RN 354-28-9

IUPAC Name: 2-chloro-2,2-difluoroacetamide
Synonyms: 2-chloro-2,2-difluoroacetamide
Molecular Weight (g/mol): 129.49
Molecular Formula: C2H2ClF2NO
InChi Key: PSAKKOKLSDIKEK-UHFFFAOYSA-N
SMILES: NC(=O)C(F)(F)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results